Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Monkey LDL ELISA Kit

Monkey LDL ELISA Kit

Monkey LDL ELISA kit
N° de catalogue ANTIA327069-96
New Product
Fournisseur:   Antibodies.com
Monkey LDL ELISA Kit
N° de catalogue ANTIA327069-96
Fournisseur:  Antibodies.com
New Product
Numéro CAS:  
Monkey LDL ELISA Kit

Monkey LDL ELISA kit

Numéro CAS:  
Prix
680,00 €
Connexion Pour la disponibilité
1 * 96 Tests
Product Details & Documents

Monkey LDL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of monkey LDL in serum, plasma, and other biological fluids.

Monkey LDL ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey LDL in serum, plasma, and other biological fluids. An antibody specific for LDL has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the LDL present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-LDL Antibody, which binds the captured LDL present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of LDL captured in each well. The concentration of LDL can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Attention: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Spécifications

Spécifications

  • Désignation:
    Monkey LDL ELISA kit
  • Type de test:
    Sandwich
  • Méthode de détection:
    Colorimetric
  • Portée de détection:
    7,81 - 500 ng/ml
  • Type déchantillon:
    Serum, plasma, and other biological fluids
  • Réactivité croisée:
    No significant cross-reactivity or interference between monkey LDL and analogues was observed.
  • Time to Results:
    3 h 30 min
  • Température de stockage:
    4 °C
  • Cat. No.:
    ANTIA327069-96
  • Format:
    Pre-coated
  • Cible:
    LDL
  • Limite de détection:
    Intra-assay CV: 4,82%, Inter-assay CV: 5,63%
  • Sensibilité:
    4,69 ng/ml
  • Durée du test:
    Multiple steps
  • Regulatory status:
    RUO
  • Durée de conservation:
    Store for 6 months at 4 °C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.