Human Apolipoprotein CII ELISA Kit
Catalog # ABCAAB168549-96
Supplier: Abcam
CAS Number:
Specifications
- Désignation:Human Apolipoprotein CII ELISA kit
- Température de stockage:+4 ℃
- Tests par set:96 tests
Specifications
About this item
Human Apolipoprotein CII ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Apolipoprotein CII in Human in plasma, serum samples.