Human Complement Factor P ELISA Kit
Catalog # ABCAAB222864-96
Fournisseur: Abcam
Numéro CAS:
Spécifications
- Désignation:Human Complement Factor P ELISA kit
- Tests par set:96 tests
Spécifications
À propos de cet article
Human Complement Factor P ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human Complement Factor P in Human in Cerebral Spinal Fluid, Saliva, Urine, plasma, serum samples.