Mouse MPO ELISA Kit (Myeloperoxidase)
Catalog # ABCAAB155458-96
Supplier: Abcam
CAS Number:
Specifications
- Désignation:Mouse MPO ELISA kit (Myeloperoxidase)
Specifications
About this item
Mouse MPO ELISA kit (Myeloperoxidase) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MPO (Myeloperoxidase) in Mouse in plasma, cell culture supernatant, cell Lysate samples.