ContactUsLinkComponent
Canine Complement C3 ELISA Kit
Canine Complement C3 ELISA Kit
Catalog # ABCAAB157697-96
Supplier:  Abcam
CAS Number:  
undefined
Canine Complement C3 ELISA Kit
Catalog # ABCAAB157697-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Désignation:
    Canine Complement C3 ELISA kit
Specifications
About this item
Canine Complement C3 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine Complement C3 in Dog in plasma, serum, Other biological fluids samples.