Human Complement H ELISA Kit
Catalog # ABCAAB213765-96
Supplier: Abcam
CAS Number:
Specifications
- Désignation:Human Complement H ELISA kit
Specifications
About this item
Human Complement H ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement H in Human in cell culture supernatant, EDTA plasma, Heparin plasma, plasma, serum, urine samples.