14,15 EET/DHET ELISA Kit
Catalog # ABCAAB175812-96
Fournisseur: Abcam
Numéro CAS:
Spécifications
- Désignation:14,15 EET / DHET ELISA kit
Spécifications
À propos de cet article
14,15 EET/DHET ELISA kit is a Competitive ELISA kit for the measurement of 14,15 EET/DHET in urine, plasma, cell culture extracts, Tissue, serum samples.