Streptavidin-HRP Solution
Streptavidin-HRP solution, 110 µl
N° de catalogue ABCAAB210901-110
Marketsource
Fournisseur: Abcam
Streptavidin-HRP Solution
N° de catalogue ABCAAB210901-110
Fournisseur: Abcam
Marketsource
Numéro CAS:
Aller à :
Product Details & Documents
Streptavidin-HRP Solution (ab210901) is part of the reagents, controls and accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.
Certifications: RUO
Spécifications
Spécifications
- Désignation:Streptavidin-HRP solution
- Taille:110 µl
- Cat. No.:ABCAAB210901-110
- Température de stockage:4 °C