MitoBiogenesis™ In-Cell ELISA Kit (Colorimetric)
Catalog # ABCAAB110217-192
Supplier: Abcam
CAS Number:
Specifications
- Durée du test:Multiple steps
- Format:Pre-coated
- Cible:MitoBiogenesis
- Désignation:MitoBiogenesis™ In-Cell ELISA kit (Colorimetric)
- Type déchantillon:Adherent cells
- Température de stockage:4 °C
- Regulatory status:RUO
- Tests par set:2×96 tests
Specifications
About this item
MitoBiogenesis™ In-Cell ELISA kit (Colorimetric) is Cell-based (quantitative) ELISA for the measurement of MitoBiogenesis™ (Colorimetric) production in Mouse, Rat, Cow, Human in Adherent cells samples.