profi ergo XG20A Safety Gloves, 60558 Series
N° de catalogue UVEX6055806
Fournisseur: UVEX
Spécifications
- Taille du gant:6
- Ambidextre/anatomique:Conception main droite et main gauche
- Usage unique/Réutilisable:Usage unique
- Poudrés/Non poudrés:Non poudrés
- Couleur:Black/orange/white
- Norme EN:EN ISO 21420:2020/EN 388:2016 + A1:2018/EN 407:2020
Spécifications
À propos de cet article
The uvex profi ergo XG safety glove range features an innovative Xtra grip coating made from nitrile rubber. It combines protection and grip with outstanding wearer comfort and flexibility.
- Xtra grip technology for outstanding grip
- Partially coated back of hand
- Excellent wearer comfort
Excellent mechanical abrasion resistance thanks to the multi-layer design for increased service life, as well as outstanding grip in damp, wet, and oily areas. Palm and ³/₄ of the back of the hand with special NBR and Xtra Grip coatings (nitrile rubber). Excellent wearer comfort due to the high water vapour absorption of the cotton lining. Free of harmful substances in accordance with OEKO-TEX® standard 100.