Aspirating pipettes
2 ml
N° de catalogue SARS86.1252.011
Marketsource
Fournisseur: SARSTEDT
Aller à :
Product Details & Documents
PS pipettes, non graduated, unplugged, sterile.
- For aspirating media with a vacuum pump
- Certified non pyrogenic
- Individually wrapped in paper plastic peel packaging
Spécifications
- Capacité:2 ml