
L(-)-Glutathion (forme oxydée) ≥98.0% (par HPLC)
Catalog # TCIAG0073-1G
Supplier: TCI
CAS Number:
Specifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-102 to -109 deg (C=2, H2O) (calcd.on anh.substance)
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:612.64 g/mol
Melting Pt:185 °C
EINECS:248-170-7
:GHS07
Storage Temperature:Réfrigérateur
CAS Number:27025-41-8
Formula:C₂₀H₃₂N₆O₁₂S₂
MDL Number:MFCD00063106