Flacons à tare
30 ml, 50×30 mm
N° de catalogue WITG2.872.005
Fournisseur: witeg Labortechnik
Aller à :
Product Details & Documents
Made of borosilicate glass.
- Excellent chemical resistance
- Excellent temperature resistance
- Permanent graduations and measures
Low form.
Spécifications
- Ø×h:50×30 mm
- Capacité:30 ml