Order Entry
Belgium
ContactUsLinkComponent
ABTS (2,2'-Azinobis[3-éthyl-2,3-dihydrobenzothiazole-6-sulfonate] de diammonium)
Irritant
ABTS (2,2'-Azinobis[3-éthyl-2,3-dihydrobenzothiazole-6-sulfonate] de diammonium)
Catalog # SERA14364.01
Supplier:  Serva
CAS Number:  
Irritant
ABTS (2,2'-Azinobis[3-éthyl-2,3-dihydrobenzothiazole-6-sulfonate] de diammonium)
Catalog # SERA14364.01
Supplier:  Serva
CAS Number:  

Specifications

  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :548.68 g/mol
EINECS:250-396-6
 :GHS07
Storage Temperature:Réfrigérateur
CAS Number:30931-67-0
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404