Dissecting scissors
Pointu/pointu, 155 mm
N° de catalogue MART11-300-15
Fournisseur: KLS Martin Group
Aller à :
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
- TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
- Straight
Spécifications
- Type:Pointu/pointu
- Longueur:155 mm