Haltegurt, Miller® TITAN™
Haltegurt, Miller® TITAN™
Safety belt, Polyamide, with 2 D-rings, 0,42 kg
N° de catalogue DALL1008232
Fournisseur:   HONEYWELL SAFETY
Haltegurt, Miller® TITAN™
N° de catalogue DALL1008232
Fournisseur:  HONEYWELL SAFETY
Haltegurt, Miller® TITAN™
Haltegurt, Miller® TITAN™
Safety belt, Polyamide, with 2 D-rings, 0,42 kg
Prix
76,10 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Safety belt for assuming the work position or for fall protection in connection with a safety harness.
  • Two light side D-rings made from stainless steel
  • Polyamide harness, width 45 mm
  • Break resistance: >15 kN
Spécifications
  • Désignation:
    Safety belt, Polyamide, with 2 D-rings
  • Poids:
    0,42 kg