Absorbants chimiques
Miniboudin M-M1002, Absorbants de maintenance, 76×2400 mm
  MMMAMM1002
 :   3M
Absorbants chimiques
  MMMAMM1002
 :  3M
Absorbants chimiques
Absorbants chimiques
Miniboudin M-M1002, Absorbants de maintenance, 76×2400 mm
168,00 €
1 * 6 ST
Chemical sorbent mini booms for laying out around equipment to contain spills and prevent leaks from spreading to neighboring work areas.
The industrial binding fleece consists mainly of inert polypropylene and polyester fibers. The lightweight and dust-free product has a high absorption capacity and reduces disposal costs to a minimum. Mini hoses for targeted containment and absorption of leaks at the base of machines.
  • For absorption of oils, coolants and non aggresive liquids
  • Booms do not float - absorb water based solutions
  • Lightweight and easy to deploy
Recommended Industries include Agriculture, Automotive, Chemicals, Construction, General Manufacturing, Healthcare, Military, Mining, Oil & Gas, Pharmaceuticals, Primary Metal, Transportation
For spill areas in chemical manufacturing, docks/piers/boats, laboratories
For spill types like acids/alkalis/alcohols, chemicals, flammable liquids, solvents - hydrocarbon chlorinated
  • Désignation:
    Miniboudin M-M1002, Absorbants de maintenance
  • Ø×L:
    76×2400 mm
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.