Working tray, benchtop, SP Bel-Art
Catalogus # BELAF246750060
Leverancier: Bel-Art Products, a Part of SP
Specificaties
- Largeur:560 mm
- Hauteur:13 mm
- Longueur:460 mm
- Cdt:1
Specificaties
Over dit artikel
Made of Polypropylene.
- Can be used in combination with biohazard, radioactive or general purpose mats
- Tray lip assures containment of spilled liquids that are not absorbed by the mats
- Autoclavable
This plastic tray is designed to hold the most popular sizes of safety mats to help contain spills.