Magnetic stirring bar retrievers, PTFE, jumbo, Spinbar®, SP Bel-Art
N° de catalogue BELAF377810000
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Ø:100 mm
- Désignation:Avec poignée en T
- Longueur:650 mm
- Cdt:1
Spécifications
À propos de cet article
Flexible PTFE magnetic retriever for removing the largest magnetic stirring bars from deep vessels. Designed with powerful Alnico 5 magnetic end having 102 mm in length.
- Easy grip 'T' handle
- Longer, custom lengths are available