Erlenmeyer flask with screw cap, WHEATON®
Erlenmeyer flask with screw cap, WHEATON®
25 ml, 20-400
N° de catalogue WHEA635134
Fournisseur:   DWK Life Sciences
Erlenmeyer flask with screw cap, WHEATON®
N° de catalogue WHEA635134
Fournisseur:  DWK Life Sciences
Erlenmeyer flask with screw cap, WHEATON®
Erlenmeyer flask with screw cap, WHEATON®
25 ml, 20-400
Prix
41,20 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These flasks, made of low extractable borosilicate glass, are used for continuous extraction of analytes from a solid into an organic solvent.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Spécifications
  • Filetage:
    20-400
  • Capacité:
    25 ml