Sampling scoops, WHEATON®
240 mm, Acier inoxydable
N° de catalogue WHEA885540
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
This scoop is made from 302 stainless steel and is ideal for sampling powders, slurries, soils, and more.
Spécifications
- Longueur:240 mm
- Matière:Acier inoxydable