Screw top vials, WHEATON®
Autosampler vial, 4,00 ml, 15×45 mm, ambré, Non
N° de catalogue WHEA224795
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
These vials made of type I borosilicate glass are designed primarily for use in the Waters WISP® 48-position and other autosamplers.
Informations de commande: Caps must be ordered separately.
Emballage: Packaged in shrink- wrapped trays of 100 for convenience and cleanliness.
Spécifications
- Avec bouchon:Non
- Ø×h:15×45 mm
- Capacité:4,00 ml
- Couleur:ambré
- Désignation:Autosampler vial