Safety floor mat for histology lab, CellMat™
Catalog # 720-2352
Supplier: CELLPATH
Specifications
- Couleur:gris
- Désignation:Safety floor mat for histology lab, CellMat X
- L×l:1000x1000 mm
- Cdt:1
Specifications
About this item
Antistatic, solvent resistant safety floor mat prevent from slipping in histology labs.
- Highly convenient and elastic
- Traps waxy dangerous deposits in its open cell type structure, which can be easily vacuumed
- Offers very good cushioning reducing fatigue to personnel who have to stand for long periods
- Edges help to anchor the mat securely to the floor and prevents from tripping
- Allows substances to be removed easily
Colour: Grey
Size: 1000×1000 mm