Potassium thiocyanate 5% en solution aqueuse pour analyses
Supplier: Bernd Kraft
Certificates
About this item
Synonyms: Potassium rhodanide
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
:97.18 g/mol
Storage Temperature:Température ambiante
EINECS:206-370-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together