2,5-Lutidine
Glass bottle for solids, 5 g
N° de catalogue MOLE78748121-5G
Marketsource
Fournisseur: Molekula
Aller à :
Product Details & Documents
Synonymes: 2,5-Dimethylpyridine
- Numéro CAS:589-93-5
- Formule:C₇H₉N
- Numéro MDL:MFCD00006343
- Poids moléculaire:107.16 g/mol
- Point d'ébullition:143 °C (1013 hPa)
- Point de fusion:–15,3 °C
- Densité:0,9346 g/cm³ (15 °C)
- EINECS:209-666-9
- UN:1993
- ADR:3,III
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:5 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



