Transport trolleys, ROLLO
Transport trolleys, ROLLO
Transport cart, ROLLO, stainless steel, 400×400×810 mm
N° de catalogue BOCH8004
Fournisseur:   BOCHEM
Transport trolleys, ROLLO
N° de catalogue BOCH8004
Fournisseur:  BOCHEM
Transport trolleys, ROLLO
Transport trolleys, ROLLO
Transport cart, ROLLO, stainless steel, 400×400×810 mm
Prix
424,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Transport cart made of 18/10 stainless steel with max. capacity of 100 kg.
Spécifications
  • Désignation:
    Transport cart, ROLLO, stainless steel
  • l×p×h:
    400×400×810 mm