Attaches, micro
0 - 20 mm, 150 mm
N° de catalogue BOCH5475
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
Micro clamps made of zinc plated steel.
- Fingers of the clamps are coated with silicone to protect clamped items
Spécifications
- Largeur de serrage:0 - 20 mm
- Longueur:150 mm