SupaMega tissue processing cassette
blanc
720-1897
: CELLPATH
CellPath’s SupaMega Cassette is ideal for embedding and processing large tissue samples. The SupaMega Cassette is widely used in research and toxicology labratories. Dimension: 65×50×15 mm
- Ideal for prostate, brain, breast tissue and eyes
- Extremely useful for research and toxicology laboratories
- Couleur:blanc