Entonnoirs de sécurité avec couvercle
Catalog # BURK2601-9900
Supplier: BURKLE
Specifications
- Filetage:2" fine thread NPT
- Cdt:1
Specifications
About this item
Stainless steel safety funnels with lid.
- Flame block
- Connector with fine thread NPT
- Discharge pipe Ø 35 mm