Gas washing bottles, DURAN®
N° de catalogue SCOT257040101
Fournisseur: DWK Life Sciences
Spécifications
- Filetage:GL 45
- Capacité:500 ml
- Environmentally Preferable:
- Cdt:1
Spécifications
À propos de cet article
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw cap system with Drechsel-head with filter disc, adjustable immersion.