Pinces Chemware®
240 mm
N° de catalogue VERND1069290
Fournisseur: Saint Gobain Life Sciences
Aller à :
Product Details & Documents
These stainless steel tongs are covered with an extra-heavy, non stick PTFE coating. For handling hot, cold, corrosive or sticky materials.
- No loss of sample through sticking
- Will not scratch or damage delicate surfaces
- Contoured jaw enables handling of crucibles and other objects
Spécifications
- Longueur:240 mm