Immersion thermostats, EH 4 basic
Fournisseur: IKA
Certificats
À propos de cet article
Immersion circulator for temperature control of up to 100 °C in open baths (with depth of at least 150 mm)
- Complies with all safety requirements for electrically operated devices
- Intended for supervised use
- Operate with non-flammable liquids only
- Suitable for all standard bath vessels
Livraison: Supplied with universal clamp.
Frequently Bought Together