Récipients à expectorations, Nunc™
Non stériles, 80 mm, Boîte de 150, 24 ml, 28,5 mm
N° de catalogue 391-7798
Fournisseur: Thermo Fisher Scientific
Récipients à expectorations, Nunc™
N° de catalogue 391-7798
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
Polystyrol.
- Low profile container can be shipped by normal mail
- Large opening facilitates secure sample placement
- Lowered centre of lid facilitates scraping off samples when the lid is removed
- Ribs on lid and container for easy grip
For sampling and transport of diagnostic specimens.
Certifications: Containers comply with 95 kPa test described in UN packaging instructions 602/650 for transport of infectious substances and diagnostic specimens.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904.
Spécifications
- Ø ext.:80 mm
- Emballage:Boîte de 150
- Type:Non stériles
- Capacité:24 ml
- Hauteur:28,5 mm
- Cat. No.:391-7798
- Certification environnementale:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact