Carnets de notes de laboratoire, Nalgene®
Carnet de notes, rouge foncé, Grille, A4, 200
N° de catalogue NALG6501-1000
Fournisseur: Thermo Fisher Scientific
Carnets de notes de laboratoire, Nalgene®
N° de catalogue NALG6501-1000
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
Polyethylene cover. Specially designed for process record keeping and ideal for research laboratories in securing patent protection. Featuring 184 high quality acid-free, paper pages with ¹/₄" grid lines. The 8¹/₂×11" pages are permanently bound and sewn.
- Page format includes place for preparer and witness signatures to aid in securing patent protection Burgundy cover is water resistant
Format: A4.
Colour: burgundy.
Spécifications
- Largeur du papier:A4
- Type:Grille
- Couleur:rouge foncé
- Désignation:Carnet de notes
- Nombre de pages:200
- Cat. No.:NALG6501-1000
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact