Order Entry
Belgium
ContactUsLinkComponent
 
Acétonitrile
Catalog # BECL290209
Supplier:  Beckman Coulter
CAS Number:  
undefined
Acétonitrile
Catalog # BECL290209
Supplier:  Beckman Coulter
CAS Number:  

Specifications

  • Type de conditionnement:
    Flacon Verre
  • Cdt:
    850 mL
  • Seq:
    2810

Specifications

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Température ambiante
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878