Stopcocks, Needle-Type, Nalgene®
Réservoirs NALGENE® jusqu'à 100 gals., 1-1/8 in.-12 straight female thread
N° de catalogue NALG6421-0010
Fournisseur: Thermo Fisher Scientific
Stopcocks, Needle-Type, Nalgene®
N° de catalogue NALG6421-0010
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
PP, white.
- Excellent chemical resistance
- Excellent temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Nalgene™ tanks up to 100 gal. with factory installed threaded boss.
Spécifications
- Pour:Réservoirs NALGENE® jusqu'à 100 gals.
- Filetage:1-1/8 in.-12 straight female thread
- Cat. No.:NALG6421-0010
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact