Potassium chlorure 3 mol/l (3 N) en solution aqueuse
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Température ambiante
EINECS:231-211-8
Frequently Bought Together