Chamber slides, Corning® BioCoat™ Poly-D-lysine/laminin
N° de catalogue 734-1072
Fournisseur: Corning
Spécifications
- Désignation:Chambre de culture, 8 puits
- Matière:PS/Verre
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:ISO 14001 - Bedford
- Cdt:12
Spécifications
À propos de cet article
These culture slides have a uniform application of poly-D-lysine/laminin.
- Tested to promote neurite outgrowth
- NG108 rat glioma/mouse neuroblastoma cells
- PS vessel lid together with removal tool