Inserts de culture cellulaire à revêtement fibronectine Corning® BioCoat™ FluoroBlok™
Catalog # 734-1053
Supplier: Corning
Specifications
- Type de membrane:PET
- Taille de pore:3,0 µm
- Désignation:Inserts de 3,0 µm dans deux plaques 24 puits
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:ISO 14001 - Bedford
- Cdt:24
Specifications
About this item
Corning® BioCoat™ FluoroBlok™ fibronectin-coated cell culture inserts packaged ready to use in 24-well Falcon® companion plates.
- Convenient, individual inserts packaged in a one-piece, automation friendly 24-multiwell format
- Combined benefits of Falcon® inserts coated with HFN for enhanced cell attachment and growth
- Individual inserts contain a 3,0 µm pore size clear PET membrane