Liqui-Nox® Phosphate-free liquid detergent
Catalog # ALCO1201
Supplier: Alconox
Specifications
- Désignation:Liqui-Nox® Phosphate-Free
- Seq:03
- Cdt:4 gal US
- Valeur pH:8,5 (for 1% solution)
- Gravité spécifique:1,07 g/ml
Specifications
About this item
Liquid detergent for cleaning analytical and EPA sampling equipment.
- Concentrated liquid detergent is phosphate-free and noncorrosive
- Completely soluble in hot or cold, hard or soft water
- Free-rinsing formula leaves no interfering residue
- Mild enough for manual cleaning (pH 8,0 to 8,5) also used in ultrasonic washers
Used in dilution of 1:100.