α-Pinène
Catalog # ROTH5647.1
Supplier: Roth Carl
CAS Number:
Specifications
- Type de conditionnement:Glass bottle
- Cdt:100 mL
- Seq:2440
Specifications
About this item
:
:136.24 g/mol
Merck Index:12,07599
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–55 °C
Density:0.8592 g/cm³ (20 °C)
EINECS:201-291-9
UN:2368
ADR:3,III
CAS Number:80-56-8
Formula:C₁₀H₁₆
MDL Number:MFCD00001339