Cristaseal
Tube sealant in plastic tray, Cristaseal
  720-0101
Cristaseal
  720-0101
Cristaseal
Cristaseal
Tube sealant in plastic tray, Cristaseal
95,10 €
1 * 10 ST
For sealing capillary tubes. The dry end of the filled tube is pressed into the Cristaseal with a twisting motion producing a firm seal with flat boundaries. The tray may be used as a carrier for tubes prior to centrifuging.
  • Quicker than flame sealing which can haemolyse the blood
  • Désignation:
    Tube sealant in plastic tray, Cristaseal