Saran Wrap
Saran™ wrap, 300 m, 450 mm
Catalog # 129-3850
Supplier: HIFI SNIJUNIE
Jump to:
Product Details & Documents
This is a total barrier film that has been specially developed for use in gel drying applications.
- Less clingy than other films, allowing it to be easily removed from gel surfaces without causing the gel to crack
Specifications
- Largeur:450 mm
- Désignation:Saran™ wrap
- Longueur:300 m
- Seq:03