Dropping funnels, cylindrical, open top, Quickfit®
Dropping funnels, cylindrical, open top, Quickfit®
50 ml, 19/26 NS, stem with cone
N° de catalogue BIBBD3/12
Fournisseur:   SciLabware
Dropping funnels, cylindrical, open top, Quickfit®
N° de catalogue BIBBD3/12
Fournisseur:  SciLabware
Dropping funnels, cylindrical, open top, Quickfit®
Dropping funnels, cylindrical, open top, Quickfit®
50 ml, 19/26 NS, stem with cone
Prix
572,00 €
Connexion Pour la disponibilité
1 * 5 ST
Product Details & Documents
PYREX®, borosilicate glass, cylindrical shape.
  • Very good chemical resistance
  • High temperature resistance
Stem inside the cone or stem with cone, open top instead of socket. Enables easy fitting to the top of chromatography cloumns etc. Easy addition of solutions. GP Rotaflo® stopcock with PTFE key for grease-free performance.
Spécifications
  • Rodage mâle:
    19/26 NS
  • Type:
    stem with cone
  • Capacité:
    50 ml