Clamps, three pronged
0 - 70 mm
N° de catalogue 241-3156
Fournisseur:   BOCHEM
Clamps, three pronged
N° de catalogue 241-3156
Fournisseur:  BOCHEM
Clamps, three pronged
Clamps, three pronged
0 - 70 mm
Prix
76,20 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Aluminium.
  • The fingers of the clamps are covered with a plastic coating to protect the items clamped
  • Without bosshead for fixing to stand
  • With T-screws screws for rapid easy fixing
Spécifications
  • Largeur de serrage:
    0 - 70 mm