Adapters, receiver, vacuum, straight, Quickfit®
201-0605
- Rodage mâle:24/29 NS
- Rodage femelle:24/29 NS
- Longueur totale:140 mm
- Environmentally Preferable:
- Cdt:1
PYREX®, borosilicate glass.
- Delivery stem within cone
- With side-arm for connection to a vacuum supply
- Easy connection to flexible tubing via detachable plastic screwthread connector