Plateaux ICSI pour FIV, Nunc™
392-0323
- Ø:51 mm
- Emballage:Conditionnement individuel
- Désignation:Plateau ICSI pour FIV, non traité, avec couvercle
- Hauteur:10 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:NAR Certificate of Retirement
- Cdt:120
Non treated surface, sterile (SAL 10⁻⁶), with lid. This IVF ICSI dish meet the exacting requirements of IVF testing, certification, and validation, providing consistently reliable, high quality results during fertilisation.
- Maintain optimal temperature with true flat bottom
- Low profile provides optimal angles during ICSI process
- Clearly see sample with super polished optical centre
- Increased safety with easy grip design
- Certificates of conformity available for every lot
Produced under ISO 13485 with full lot traceability
: Produced under ISO 13485 with full lot traceability.