Millicell® EZ Slide Holder
Catalog # MILFPEZXMSH01
Supplier: Merck
Specifications
- Désignation:Microscope slide holder for 4 and 8 well units, holds two slides, L×W×H: 128×86×147/17/95 mm, black
- Matière:PS
- Seq:05
- Cdt:1
Specifications
About this item
Microscope slide holder for 4 and 8 well units, holds two slides.
- Shipped in ambient temperature
Techniques: cell based assay: suitable cell culture; mammalian: suitable immunocytochemistry: suitable