Specifications
- Couleur:vert/gris
- Désignation:Planet safety spectacle
- Oculaires:Clear
- Cdt:1
Specifications
About this item
Attractive, classic, sporty design, pleasantly lightweight and optimal protection - the uvex pheos nxt. The special design of the arms not only looks impressive: the direction-focused structure also provides extremely good grip - even during demanding tasks. Numerous openings in the side arms improve air circulation and make the glasses light yet no less robust. The large panoramic lens also provides a wide field of vision free from distortion to ensure a good view.
- Classic, sporty and lightweight with innovative arm design for an optimal, reliable fit
- uvex protecting planet - mass-balanced PC lens, arms and packaging partially made from recycled material
- Robust polycarbonate lens with proven uvex supravision coating technology - permanently anti-fog and scratch-resistant lens
- Metal-free
- Frame colour: Anthracite, jade
- PC lens: clear
- UV400 (UV protection)
- Anti-fog on the inside
- Extremely scratch-resistant
- Chemical-resistant on the outside
Ergonomically shaped arms with grip structure and soft ear-pieces ensure a comfortable and secure fit without creating pressure points. Increased air circulation for outstanding, climate-optimised wearer comfort through numerous openings in the side arms. Anatomically shaped nose piece for all-day wearer comfort.