Human Leukopak
Fournisseur:   PROVITRO
Human Leukopak
Fournisseur:  PROVITRO
Certificats
Human Leukopak
10 Options disponibles ci-dessous
Product Details & Documents
Our highly cell-concentrated Leukopak is a specialised product from leukapheresis using the Spectra Optia system. This advanced technology employs centrifugal mechanisms for continuous blood flow, extracting white blood cells from peripheral blood into a sterile bag with ACD-A anticoagulant. The enriched product includes monocytes, lymphocytes, platelets, plasma, and erythrocytes.
  • Leukopak is collected from healthy donors using approved permission forms and procedures
  • Collection procedures have been approved by the Institutional Review Board (IRB)
  • The product includes donor information and serology testing for: HIV, HBs-AG, HCV
  • Additional optional information and testing are available for a small fee
Our offerings encompass Leukopaks sourced from our existing inventory, alongside the provision for prospective collections tailored to specific research inquiries. This versatile range ensures a breadth of options to suit a variety of research requirements, whether drawing from our readily available stock or arranging for custom collections based on unique project needs.
Storage: Prepare cells for extended storage in the vapor phase of liquid nitrogen or thaw them for immediate use. Avoid storing cells in the liquid phase of nitrogen. Short-term storage (less than two weeks) at −80 °C is acceptable but should be minimised for optimal stability. Once thawed, it's crucial to use the samples immediately.
 : Not intended for any human or animal diagnostic or therapeutic use.
Product Family Options
Informations sur le produit
  • Désignation
    Format de cellule
    Donneur unique/donneurs regroupés
    Cdt
    Disponibilité
    Prix
  • Human leukopak, healthy donor, 2,5B cells (2,5×10⁹ cells/bag)
    Fresh
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 3 520,00 €
    3 520,00 €
    1 * 1 ST
       3 520,00 €
  • Human leukopak, healthy donor, 5B cells (5×10⁹ cells/bag)
    Fresh
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 4 820,00 €
    4 820,00 €
    1 * 1 ST
       4 820,00 €
  • Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag)
    Fresh
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 5 480,00 €
    5 480,00 €
    1 * 1 ST
       5 480,00 €
  • Human leukopak, healthy donor, 20B cells (20×10⁹ cells/bag)
    Fresh
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 8 220,00 €
    8 220,00 €
    1 * 1 ST
       8 220,00 €
  • Human leukopak, diseased donor
    Cryopreserved
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST
  • Human leukopak, healthy donor, 2,5B cells (2,5×10⁹ cells/bag)
    Cryopreserved
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 3 270,00 €
    3 270,00 €
    1 * 1 ST
       3 270,00 €
  • Human leukopak, healthy donor, 5B cells (5×10⁹ cells/bag)
    Cryopreserved
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 4 360,00 €
    4 360,00 €
    1 * 1 ST
       4 360,00 €
  • Human leukopak, healthy donor, 10B cells (10×10⁹ cells/bag)
    Cryopreserved
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 5 130,00 €
    5 130,00 €
    1 * 1 ST
       5 130,00 €
  • Human leukopak, healthy donor, 20B cells (20×10⁹ cells/bag)
    Cryopreserved
    Single donor
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST - 7 900,00 €
    7 900,00 €
    1 * 1 ST
       7 900,00 €
  • Human leukopak, diseased donor
    Fresh
    Single donor